Skip to Content
Merck
All Photos(1)

Key Documents

AB1623

Sigma-Aldrich

Anti-Histone H2B Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$824.00

$824.00


Estimated to ship on24 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1624

Request a Bulk Order

Select a Size

Change View
100 μL
$824.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$824.00


Estimated to ship on24 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1624

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... H2BC1(255626)

Specificity

Reacts with histone H2B in human nuclear fractions. No cross-reactivity with other histones, H1, H2A, H3 or H4. Binding of histones from other species has not been tested, but it is likely as the amino acid sequence of histone H2B has been found to be identical in all species examined.

Immunogen

Synthetic peptide

Application

Detect Histone H2B also known as Histone H2B with Anti-Histone H2B Antibody (Rabbit Polyclonal Antibody), that has been demonstrated to work in WB & IHC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Western blot

Immunohistochemistry on paraformaldehyde treated sections.

Optimal working dilutions must be determined by end user.

Linkage

Replaces: MABE15

Physical form

Rabbit antiserum. Liquid. Contains no preservative.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siwei Ouyang et al.
Neurobiology of aging, 36(10), 2850-2860 (2015-08-11)
The progressive loss of neurons causes neurodegenerative diseases. Because the accumulation of DNA breaks results in neuronal apoptosis, the lack of a variety of DNA damage repair-related proteins contributes to neurodegeneration. The ubiquitin ligase RNF8 plays an important role in
Ricardo Martinez-Zamudio et al.
Molecular and cellular biology, 32(13), 2490-2502 (2012-05-02)
The packaging of DNA into nucleosomes imposes obstacles on gene transcription, and histone-modifying and nucleosome-remodeling complexes work in concert to alleviate these obstacles so as to facilitate transcription. Emerging evidence shows that chromatin-associated poly(ADP-ribose) polymerase 1 (PARP-1) and its enzymatic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service