Skip to Content
Merck
All Photos(1)

Key Documents

AB1587P

Sigma-Aldrich

Anti-Adenosine A1 Receptor Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$964.00

$964.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$964.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$964.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat, sheep, guinea pig, pig

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ADORA1(134)

Specificity

Recognizes Adenosine A1 receptor (A1R). Recognizes the 79 kDa and 39 kDa bands of in cortical membranes from rat, porcine and lamb. The 79 kDa band can be converted to 39 kDa band by agonist or antagonist treatment and could represent a A1R dimers.

Immunogen

Amino acid sequence corresponding to the 3rd extracellular domain (amino acids 163-176, GEPVIKCEFEKVIS) of the rat A1R gene.

Application

Anti-Adenosine A1 Receptor Antibody detects level of Adenosine A1 Receptor & has been published & validated for use in ELISA, IC, IH & WB.
Immunohistochemistry: 10-50 μg/mL on frozen tissue sections. Light fixation is recommended. 2% 0.2% picric acid {Cooke, HJ (1999) AJP Gastrointest Liver Physiol 276:4516-4626}

Immunocytochemistry: Caufield, 2003. Light 2% PFA fixation 5-10 minutes --long fixations lower staining intensity.

Western blot: 1-10 μg/mL (Chemiluminescence technique): Membrane preparations are recommended for enhanced signals {Smith et al (2001) Biochem Pharmacology 61:237-244} Higher concentrations of the antibody may be necessary if calorimetric detection or milk-based antibody diluent are used.

ELISA: 1:10,000-1:50,000 (50-100 ng A1R peptide (Cat. Number AG289)/well)

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Circadian Rhythm & Sleep

Target description

79 kDa and 39 kDa

Physical form

Affinity Purified immunoglobulin. Liquid in PBS with 0.1% BSA.
ImmunoAffinity Purified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cátia Vieira et al.
Mediators of inflammation, 2014, 254640-254640 (2014-09-12)
Purinergic signalling is remarkably plastic during gastrointestinal inflammation. Thus, selective drugs targeting the "purinome" may be helpful for inflammatory gastrointestinal diseases. The myenteric neuromuscular transmission of healthy individuals is fine-tuned and controlled by adenosine acting on A(2A) excitatory receptors. Here
Patrycja Bolewska et al.
Neuroscience journal, 2019, 5950818-5950818 (2019-07-06)
There is an excellent correlation between the age when alcohol consumption begins and the likelihood of lifelong problems with alcohol abuse. Alcohol use often begins in adolescence, a time marked by brain development and maturation of numerous brain systems. Rats
Carina Herman-de-Sousa et al.
Cells, 9(3) (2020-03-12)
Human subcutaneous fibroblasts (HSCF) challenged with inflammatory mediators release huge amounts of ATP, which rapidly generates adenosine. Given the nucleoside's putative relevance in wound healing, dermal fibrosis, and myofascial pain, we investigated the role of its precursor, AMP, and of
Ella E Bauer et al.
Neural plasticity, 2020, 5859098-5859098 (2020-05-14)
Adenosine acts as a key regulator of striatum activity, in part, through the antagonistic modulation of dopamine activity. Exercise can increase adenosine activity in the brain, which may impair dopaminergic functions in the striatum. Therefore, long-term repeated bouts of exercise

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service