Skip to Content
Merck
All Photos(1)

Documents

AB1551

Sigma-Aldrich

Anti-Metabotropic Glutamate Receptor 1α Antibody, pain, CT

Chemicon®, from rabbit

Synonym(s):

mGluR1alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, monkey, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GRM1(2911)

Specificity

Recognizes metabotropic glutamate receptor 1a (mGluR1a). On western blots of rat brain microsomes, the antibody labels a single band migrating at 140 kDa, corresponding to mGluR1alpha (Testa, 1998). The antibody recognizes a 140 kDa band in extracts from transfected cells expressing mGluR1alpha but not in extracts from transfected cells expressing mGluR5 (Marino, 2001).

Immunogen

Carboxy terminal peptide of rat mGluR1alpha conjugated to KLH with glutaraldehyde (PNVTYASVILRDYKQSSSTL).
Epitope: C-terminus

Application

Anti-Metabotropic Glutamate Receptor 1α Antibody, pain, C-terminus is an antibody against Metabotropic Glutamate Receptor 1α for use in IH(P), IH & WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot: 0.25-1.0 μg/mL. Detects a band migrating at approximately 140 kDa. The use of membrane preparations and protease inhibitors is highly recommended.

Immunohistochemistry: AB1551 can be used for immunocytochemistry using paraformaldehyde or paraformaldehyde/glutaraldehyde fixed tissue with light and electron microscopy. Cryostat and vibratome sections can be used with or without Triton X-100 treatment. Final concentrations recommended are 0.5-2.0 μg/mL.

The antibody has not been tested for use in immunoprecipitation. However, antibodies to the carboxy termini of other glutamate receptors are very effective in immunoprecipitation.

Optimal working dilutions must be determined by end user.

Target description

140 kDa

Linkage

Replaces: AB1595

Physical form

Affinity Purified using the immunogen peptide. Lyophilized from PBS containing 1% BSA.
ImmunoAffinity Purified

Storage and Stability

Maintain lyophilized material at 2–8°C or -20°C. Reconstitute to 500 μL with sterile distilled water. Store reconstituted material frozen (-20°C or -80°C) in undiluted aliquots for up to six months. Avoid repeated thawing and refreezing. Diluted antibody containing 0.1% sodium azide for Western blot analysis can be stored for several weeks and reused repeatedly.

Analysis Note

Control
Cerebellar tissue sections or homogenates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Up-regulation of hippocampal metabotropic glutamate receptor 5 in temporal lobe epilepsy patients.
Notenboom, RG; Hampson, DR; Jansen, GH; van Rijen, PC; van Veelen, CW; van Nieuwenhuizen et al.
Brain null
Co-localization of caldesmon and calponin with cortical afferents, metabotropic glutamate and neurotrophic receptors in the lateral and central nuclei of the amygdala.
Agassandian, K; Cassell, MD
Brain Research null
Ultrastructural relationships between cortical, thalamic, and amygdala glutamatergic inputs and group I metabotropic glutamate receptors in the rat accumbens.
Mitrano, DA; Pare, JF; Smith, Y
The Journal of Comparative Neurology null
D A Mitrano et al.
Neuroscience, 154(2), 653-666 (2008-05-16)
There is significant pharmacological and behavioral evidence that group I metabotropic glutamate receptors (mGluR1a and mGluR5) in the nucleus accumbens play an important role in the neurochemical and pathophysiological mechanisms that underlie addiction to psychostimulants. To further address this issue
Chirag Upreti et al.
Scientific reports, 11(1), 5552-5552 (2021-03-12)
Retinoid X receptors are members of the nuclear receptor family that regulate gene expression in response to retinoic acid and related ligands. Group 1 metabotropic glutamate receptors are G-protein coupled transmembrane receptors that activate intracellular signaling cascades in response to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service