Skip to Content
Merck
All Photos(1)

Key Documents

AB15506

Sigma-Aldrich

Anti-Iron Pegulatory Protein 1 Antibody

Chemicon®, from rabbit

Synonym(s):

IRP-1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$556.00

$556.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$556.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$556.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ACO1(48)

General description

Iron regulatory proteins (IRP-1 and IRP-2) are cytoplasmic mRNA-binding proteins that control intracellular iron levels by regulating the translation of ferritin, Tfrs, and other proteins congaing iron-responsive element (IRE) located at the 5′-UTR. IRP binds to IREs with high affinity in situation of iron starvation. Binding of IRP to IRE of ferritin represses its transcription. However, when cells are iron replete, IRPs lose their capacity to bind IREs, allowing efficient translation of ferritin and reducing Tfr mRNA half-life. Although IRP1 and IRP-2 share significant protein sequence homology, they differ in tissue distribution and mode of regulation. IRP-2/IRE-BP2 (rat/human 963 aa) is relatively less abundant than IRP-1 and lacks aconitase activity. It has a unique 73-aa insertion in the N-terminus. In the presence of high iron levels, IRP-2 is rapidly targeted to proteasome-mediated degradation. Although, both IRPs bind the consensus IREs, it is also shown that IRP-2 can recognize an exclusive IRE subset.

Specificity

Iron regulatory protein 1 (IRP-1). The immunogen sequence has no significant homology with IRP-2.
Other species have not yet been tested. The immunogen sequence is 94% conserved in mouse, and 89% in human and rabbit IRP-1.

Immunogen

Synthetic peptide near the N-terminus of rat IRP-1.

Application

Anti-Iron Pegulatory Protein 1 Antibody detects level of Iron Pegulatory Protein 1 & has been published & validated for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Neurofilament & Neuron Metabolism

RNA Binding Protein (RBP)
Western blot: 1-10 µg/mL using ECL.
Immunohistochemistry: Not tested. Recommended testing antibody dilution:2-20 µg/mL
ELISA: 1:10,000-1:100,000 using 50-100 ng of control peptide/well.
Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
ImmunoAffinity Purified
Liquid in PBS with 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

? -Actinin TvACTN3 of Trichomonas vaginalis is an RNA-binding protein that could participate in its posttranscriptional iron regulatory mechanism.
Calla-Choque, JS; Figueroa-Angulo, EE; Avila-Gonzalez, L; Arroyo, R
BioMed Research International null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service