Skip to Content
Merck
All Photos(1)

Documents

AB1534SP

Sigma-Aldrich

Anti-Brain Derived Neurotrophic Factor Antibody

Chemicon®, from rabbit

Synonym(s):

BDNF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... BDNF(627)

Specificity

Recognizes Brain Derived Neurotrophic Factor (BDNF). By dot blot, less than 0.1% cross-reactivity against mouse NGF, recombinant human NT3 or neurotrophin 4.

Immunogen

A 10 amino acid peptide corresponding to the amino-terminal of mouse Brain Derived Neurotrophic Factor (BDNF) coupled to ovalbumin. The sequence is H2N-HSDPARRGEL-COOH.

Application

Anti-Brain Derived Neurotrophic Factor Antibody is an antibody against Brain Derived Neurotrophic Factor for use in ELISA, IH & WB.
Immunohistochemistry: 1-10 μg/mL (see suggested protocol).

Immunoblotting: 1-10 μg/mL. BDNF is not easily extracted in standard buffers. Dissected tissues should be homogenized in 10 volumes of 100mM phosphate buffer containing 1mM EDTA, 2M guanidine hydrochloride (pH7.2), and three protease inhibitors, 10mM N-ethylmaleimide, 0.36mM pepstatin, and 1mM PMSF. Homogenates are then sonicated and centrifuged at 46K x g for 30 minutes at 4°C. BDNF migrates as 18 kDa, with a homodimer at ~27-30 kDa.

ELISA (one-site): 1-10 μg/mL

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Neuroinflammation & Pain

Target description

18 kDa (monomer) and 27-30 kDa (homodimer)

Physical form

Format: Purified
Protein A Purified immunoglobulin fraction lyophilized from PBS, with no preservatives. Reconstitute with 500 μL of sterile distilled water (1 mg/mL).
Protein A purified

Storage and Stability

Maintain at -20 to -70°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. Glycerol (1:1) can be added for additional stability.

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonino Germanà et al.
International journal of molecular sciences, 21(16) (2020-08-19)
Brain-derived neurotrophic factor (BDNF), a member of the neurotrophin family, is involved in multiple and fundamental functions of the central and peripheral nervous systems including sensory organs. Despite recent advances in knowledge on the functional significance of BDNF and TrkB
Luciano Domenici et al.
PloS one, 9(12), e115579-e115579 (2014-12-24)
Vision loss in glaucoma is caused by progressive dysfunction of retinal ganglion cells (RGCs) and optic nerve atrophy. Here, we investigated the effectiveness of BDNF treatment to preserve vision in a glaucoma experimental model. As an established experimental model, we
Ewa Badowska-Szalewska et al.
Acta neurobiologiae experimentalis, 77(1), 68-76 (2017-04-06)
The way hippocampal neurons function during stress in old age (critical times of life) is dependent on brain derived neurotrophin factor (BDNF). This study examined the influence of acute and chronic forced swim (FS) or high-light open field (HL‑OF) stimulation
Marialuisa Aragona et al.
International journal of molecular sciences, 23(9) (2022-05-15)
Brain-derived neurotrophic factor (BDNF) is a member of the neurotrophin family and it is involved in several fundamental functions in the central and peripheral nervous systems, and in sensory organs. BDNF regulates the chemosensory systems of mammals and is consistently
Anam Hammid et al.
Drug metabolism and disposition: the biological fate of chemicals, 50(12), 1483-1492 (2022-10-05)
As a multitissue organ, the eye possesses unique anatomy and physiology, including differential expression of drug-metabolizing enzymes. Several hydrolytic enzymes that play a major role in drug metabolism and bioactivation of prodrugs have been detected in ocular tissues, but data

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service