Recommended Products
biological source
goat
Quality Level
antibody form
purified antibody
clone
polyclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable
shipped in
wet ice
Specificity
Influenza B, purified virions. Non reactive with monkey kidney cells.
No cross-reactivity with Influenza A virus, Parainfluenza virus (types 1-3), RSV or Adenovirus.
No cross-reactivity with Influenza A virus, Parainfluenza virus (types 1-3), RSV or Adenovirus.
Immunogen
Yamagata strain of Influenza B
Application
Detect Influenza B using this Anti-Influenza B Antibody validated for use in ELISA, IF, WB, IH.
ELISA
Fluorescence microscopy
Immunoblotting; unboiled samples in 1mM DTT and 2% SDS only.
Immunohistochemistry.
IFA: >1:1,000.
Optimal working dilutions must be determined by end user.
Fluorescence microscopy
Immunoblotting; unboiled samples in 1mM DTT and 2% SDS only.
Immunohistochemistry.
IFA: >1:1,000.
Optimal working dilutions must be determined by end user.
Research Category
Infectious Diseases
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
Infectious Diseases - Viral
Physical form
Format: Purified
IgG fraction. Liquid in PBS (0.01 M, pH 7.2) and 0.1% sodium azide. >95% pure.
Storage and Stability
Maintain at +2-8°C for 3 months or at -20°C in aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. Store under subdued light.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of allergy and clinical immunology, 139(5), 1575-1582 (2017-02-18)
Antibody responses to the inactivated seasonal influenza vaccine in patients with atopic dermatitis (AD) have not been carefully characterized. The primary objective of this study was to compare antibody responses to intradermal vaccination in participants with moderate/severe AD with those
Bral1: its role in diffusion barrier formation and conduction velocity in the CNS.
The Journal of Neuroscience null
Brevican distinctively assembles extracellular components at the large diameter nodes of Ranvier in the CNS.
Journal of Neurochemistry null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service