Skip to Content
Merck
All Photos(2)

Documents

AB10526

Sigma-Aldrich

Anti-COX4 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

cytochrome c oxidase subunit IV isoform 1, cytochrome c oxidase subunit IV1, Cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1, mitochondrial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... COX4I1(1327)

Related Categories

General description

Cytochrome C oxidase subunit 4 (COX4), is one of 13 subunits of cytochrome c oxidase. Cytochrome c oxidase is responsible for catalyzing the last step of electron transfer through the respiratory chain on the mitochondrial inner membrane. COX4 assembles into sub-complexes with other subunits of cytochrome c oxidase and may represent a rate-limiting intermediate.

Specificity

Other homologies: Rat and mouse (59% sequence homology).

Immunogen

KLH-conjugated linear peptide corresponding to human COX4.

Application

Anti-COX4 Antibody detects level of COX4 & has been published & validated for use in WB, IH(P).
Immunohistochemistry Analysis: A 1:500 dilution from a previous lot detected COX4 in normal human cardiac muscle tissue.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blot in human fetal skeletal muscle tissue lysate.

Western Blot Analysis: 0.1 µg/mL of this antibody detected COX4 in 10 µg of human fetal skeletal muscle tissue lysate.

Target description

~17 kDa observed.
A non-specific band may be observed at ~58 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human fetal skeletal muscle tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shekoufeh Almasi et al.
The Journal of biological chemistry, 293(10), 3637-3650 (2018-01-19)
A lack of effective treatment is one of the main factors contributing to gastric cancer-related death. Discovering effective targets and understanding their underlying anti-cancer mechanism are key to achieving the best response to treatment and to limiting side effects. Although
Ana Andres-Hernando et al.
American journal of physiology. Renal physiology, 317(4), F941-F948 (2019-08-15)
Obesity and metabolic syndrome are well-known risk factors for chronic kidney disease (CKD); however, less is known about the mechanism(s) by which metabolic syndrome might accelerate kidney disease. We hypothesized that metabolic syndrome should accelerate the development of kidney disease

Articles

Loading controls in western blotting application.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service