Skip to Content
Merck
All Photos(1)

Key Documents

AB10200

Sigma-Aldrich

Anti-Perilipin A Antibody

Chemicon®, from rabbit

Synonym(s):

Anti-FPLD4, Anti-PERI, Anti-PLIN

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$214.00

$214.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$214.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$214.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

bovine, mouse

packaging

antibody small pack of 25 μg

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Plin1(103968)

General description

Perilipin A is the predominant isoform of perilipins a family of proteins which are the most abundant proteins at the surfaces of lipid droplets in adipocytes and are also found in steroidogenic cells.

Immunogen

Bovine Perilipin A

Application

Suggested dilutions:
Western Blot: 0.1-0.2 μg/mL
Detect Perilipin A using this Anti-Perilipin A Antibody validated for use in WB.
Research Category
Metabolism
Research Sub Category
Lipid Metabolism & Weight Regulation

Quality

Routinely evaluated by immunoblot.

Target description

64kDa

Physical form

Format: Purified
Purified immunoglobulin presented as a liquid in 0.1 M Na-Phosphate buffer, pH 7.0 containing protease free bovine serum albumin.

Storage and Stability

Maintain frozen at -20C in undiluted aliquots for up to 12 months from date of receipt. Avoid repeated freeze-thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophia J Erb et al.
Scientific reports, 12(1), 888-888 (2022-01-20)
Knockdown of patatin-like phospholipase domain-containing protein 3 (PNPLA3) increased triglycerides (TG) in primary bovine hepatocytes, suggesting that PNPLA3 plays a causal role in hepatic TG clearing. In vivo, PNPLA3 abundance across the periparturient period is inversely related to hepatic TG
Charles K F Chan et al.
Cell, 175(1), 43-56 (2018-09-23)
Stem cell regulation and hierarchical organization of human skeletal progenitors remain largely unexplored. Here, we report the isolation of a self-renewing and multipotent human skeletal stem cell (hSSC) that generates progenitors of bone, cartilage, and stroma, but not fat. Self-renewing and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service