Skip to Content
Merck
All Photos(2)

Key Documents

AB10029

Sigma-Aldrich

Anti-Ubiquityl Histone H2A.X (Lys119) Antibody

serum, from rabbit

Synonym(s):

H2AXK119Ub, Histone H2A.X Ubiquityl K119), H2A histone family, member X, H2AX histone

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$910.00

$910.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$910.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$910.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), bovine (based on 100% sequence homology), canine (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

ubiquitination (Lys119)

Gene Information

bovine ... H2Ax(531733)
dog ... H2Ax(489372)
human ... H2AX(3014)
mouse ... H2Ax(15270)
rat ... H2Ax(500987)

General description

H2AX is involved in DNA repair and the maintenance of genomic stability. It is been implicated both in homologous recombination and nonhomologous end joining DNA repair pathways. H2AX is thought to have a critical function in the recruitment of DNA repair factors and DNA damage-signaling proteins. H2A.X is ubiquitinated by RING1 and RNF2/RING2 complex providing a specific mark for transcriptional repression. In addition this protein can be ubiquitinated by the E2 ligase UBE2N and the E3 ligases RNF8 and RNF168 as result of double-stranded DNA breaks to recruit repair proteins a damaged site. H2AX is phosphorylated soon after the appearance of double-stranded breaks. Hyperphoshorylation of H2AX may be linked to chromatin fragmentation prior to apoptosis.


Specificity

This antibody recognizes Histone H2A.X when ubiquitylated at Lys119.

Immunogen

Epitope: Lys119
Linear peptide containing ubiquitylated Lys119 of human Histone H2A.X.

Application

Anti-Ubiquityl Histone H2A.X (Lys119) Antibody is a rabbit polyclonal antibody for detection of Ubiquityl Histone H2A.X (Lys119) also known as H2AXK119Ub, Histone H2A.X Ubiquityl K119) & has been validated in WB, IP, ICC.
Immunocytochemsitry Analysis: 1:500 dilution from a representative lot detected Histone H2A.X in RAW264.7 and NIH/3T3 cells.

Immunoprecipitation Analysis: 5 µl from a representative lot immunoprecipitated Histone H2A.X in HeLa acid extract.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Evaluated by Western Blot in HeLa acid extract.

Western Blot Analysis: 1:1,000 dilution of this antibody detected Histone H2A.X on 10 µg of HeLa acid extract.

Target description

~ 23 kDa

Physical form

Unpurified
Unpurified rabbit polyclonal containing serum with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Analysis Note

Control
HeLa acid extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xing-Xiao Yang et al.
Oncology reports, 39(2), 667-678 (2017-12-06)
B-cell‑specific Moloney murine leukaemia virus integration site-1 (BMI-1) contributes to the growth of tumour cells post-irradiation (IR). The aim of the present study was to characterize the effects of BMI-1 on cell viability, radiosensitivity and its mechanisms of action in
Stéphanie Olivier-Van Stichelen et al.
Frontiers in genetics, 5, 256-256 (2014-08-20)
O-GlcNAc Transferase (OGT) catalyzes protein O-GlcNAcylation, an abundant and dynamic nuclear and cytosolic modification linked to epigenetic regulation of gene expression. The steady-state levels of O-GlcNAc are influenced by extracellular glucose concentrations suggesting that O-GlcNAcylation may serve as a metabolic
Yannick D Benoit et al.
Journal of cellular physiology, 228(4), 764-772 (2012-09-25)
Colorectal cancer is ranked among the top leading causes of cancer death in industrialized populations. Polycomb group proteins, including Suz12 and Ezh2, are epigenetic regulatory proteins that act as transcriptional repressors of many differentiation-associated genes and are overexpressed in a
Nicole Kaminski et al.
Molecular cell, 82(21), 4001-4017 (2022-10-21)
Alternative lengthening of telomeres (ALT) is a homology-directed repair (HDR) mechanism of telomere elongation that controls proliferation in subsets of aggressive cancer. Recent studies have revealed that telomere repeat-containing RNA (TERRA) promotes ALT-associated HDR (ALT-HDR). Here, we report that RAD51AP1
Sivaprakasam Balasubramanian et al.
Cellular signalling, 27(7), 1336-1344 (2015-04-07)
The Bmi-1 Polycomb group (PcG) protein is an important epigenetic regulator of chromatin status. Elevated Bmi-1 expression is observed in skin cancer and contributes to cancer cell survival. (-) Epigallocatechin-3-gallate (EGCG), an important green tea-derived cancer prevention agent, reduces Bmi-1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service