Skip to Content
Merck
All Photos(1)

Key Documents

960212

Supelco

Filling nozzle (stainless steel)

PTFE tube, stainless steel coating, tube L 80 cm, self-closing, for rapid action connector

Synonym(s):

Filling nozzle with tubing

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$6,130.00

$6,130.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$6,130.00

About This Item

UNSPSC Code:
41121516
NACRES:
NB.51

$6,130.00


Estimated to ship on28 May 2025


Request a Bulk Order

material

PTFE tube
stainless steel coating

feature

self-closing

tube L

80 cm

fitting

for rapid action connector

shipped in

ambient

storage temp.

room temp

Related Categories

General description

Different environments and individual customer applications require special withdrawal options. Optimized nozzles are available for each special application, e.g. withdrawal inside a fume hood with a flexible filling nozzle while the barrel is installed inside a safety cabinet. This filling nozzle consists of 9.67100.1090 Filling nozzle and 9.67100.9052 Stainless steel coated PTFE-tube, length 80 cm.
self-closing, with stainless steel-coated PTFE-tube (80 cm) and rapid action connector

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service