Skip to Content
Merck
All Photos(1)

Documents

9081-M

Sigma-Aldrich

Human Leptin

Synonym(s):

Leptin hormone

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405

biological source

human

Quality Level

label

125Iodine labeled

form

solid

NCBI accession no.

UniProt accession no.

Gene Information

human ... LEP(3952)

General description

Radioactivity: <3 μCi
These iodinated hormonos are HPLC purified and prepared fresh monthly.

Application

appropriate label hydrating buffer if kit replacement or assay buffer of choice

Storage and Stability

2–8 °C

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service