8800-OP
TBE Buffer
OmniPur® Grade, 5X Liquid Concentrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105319
Recommended Products
General description
Each liter of concentrate makes 5L of 1X TBE. Molecular biology grade.
Specifications:
Conductivity 20%/H2O @ 25°C: 1065 µmho max
DNase: None detected
pH (20%, water): at 25°C: 8.2-8.4
pH of solution at 25°C: report
Protease: None detected
RNase: None detected
Titration Total in 5X Tris + Boric acid + EDTA: 0.875-0.965 M
Intended for laboratory and manufacturing use only. Not for drug, food, or household use.
Specifications:
Conductivity 20%/H2O @ 25°C: 1065 µmho max
DNase: None detected
pH (20%, water): at 25°C: 8.2-8.4
pH of solution at 25°C: report
Protease: None detected
RNase: None detected
Titration Total in 5X Tris + Boric acid + EDTA: 0.875-0.965 M
Intended for laboratory and manufacturing use only. Not for drug, food, or household use.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service