Skip to Content
Merck
All Photos(1)

Key Documents

8.55063

Sigma-Aldrich

NovaSyn TG bromo resin

Novabiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$59.70
5 G
$206.00

$59.70


0 left in stock

Request a Bulk Order

Select a Size

Change View
1 G
$59.70
5 G
$206.00

About This Item

UNSPSC Code:
12352101
NACRES:
NA.22

$59.70


0 left in stock

Request a Bulk Order

Quality Level

product line

NovaSyn® TG
Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

General description

NovaSyn TG bromo resin is a composite of low cross-linked polystyrene and 3000-4000 M.W. polyethylene glycol [1,2,3], in which the ends of the PEG chains have been bromo functionalized. The 130 μm beads have a narrow size distribution, high diffusion rates and excellent swelling properties across a wide range of solvents from toluene to water, making them ideally suited to the synthesis of serial and parallel libraries by organic solid phase synthesis. This support may be used for the immobilization of alcohols, carboxylic acids, phenols and thiols, or as base support for other linkers [4,5]. Loading of this support can be carried out using the same methods as those previously described for Merrifield resin.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] W. Rapp, et al. in ′Innovation & Perspectives in Solid Phase Synthesis, 1st International Symposium′, R. Epton (Eds), SPCC UK Ltd., Birmingham, 1990, pp. 205.
[2] L. Zhang, et al. in ′Peptides 1990, Proc. 21st European Peptide Symposium′, E. Giralt & D. Andreu (Eds), ESCOM, Leiden, 1990, pp.196.
[3] E. Bayer (1991) Angew. Chem. Int. Ed. Engl., 30, 113.
[4] K. H. Gordon & S. Balasubramanian (2001) Org. Lett., 3, 53.
[5] D. Tumelty, et al. (2001) Org. Lett., 3, 83

Linkage

Replaces: 01-64-0095

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined by elemntal analysis of bromine): 0.20 - 0.40 mmol/g
Swelling Volume (in CH₂Cl₂): lot specific result

Legal Information

NOVASYN is a registered trademark of Merck KGaA, Darmstadt, Germany
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service