Skip to Content
Merck
All Photos(1)

Key Documents

8.55026

Sigma-Aldrich

Wang resin (100-200 mesh)

Novabiochem®

Synonym(s):

Aldehyde Wang HL resin

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$143.00
25 G
$575.00
100 G
$1,720.00

$143.00


Only 0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$143.00
25 G
$575.00
100 G
$1,720.00

About This Item

UNSPSC Code:
13111023
NACRES:
NA.22

$143.00


Only 0 left in stock

Request a Bulk Order

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

alcohol

storage temp.

2-8°C

General description

This support is an excellent tool for the production of sulfonamides and anilides by SPOS [1]. Amines have been released from this resin with DDQ [2] and tertiary amides by acylation of resin-bound secondary amines with acid chlorides [3].

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] B. Raju & T. P. Kogan (1997) Tetrahedron Lett., 38, 4965.
[2] S. Kobayashi & Y. Aoki (1998) Tetrahedron Lett., 39, 7345.
[3] M. W. Miller, et al. (1998) Tetrahedron Lett., 39, 3429.

Linkage

Replaces: 01-64-0182

Analysis Note

Color (visual): white to yellow to orange
Appearance of substance (visual): beads
Loading (determined by elemental analysis of sulfur after derivatization with tosylhydrazide): 2.0 - 3.5 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly(styrene - 2% DVB), 200 -400 mesh.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Good Afternoon, in which organic solvent Is this resin soluble? Thanks

    1 answer
    1. The recommended solvent to swell this resin is Dimethylformamide.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service