Skip to Content
Merck
All Photos(1)

Key Documents

8.52338

Sigma-Aldrich

Fmoc-thioproline

Novabiochem®

Synonym(s):

Fmoc-thioproline, Fmoc-thiaproline,Fmoc-thiazolidine-4-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$50.30
5 G
$201.00
25 G
$793.00

$50.30


0 left in stock

Request a Bulk Order

Select a Size

Change View
1 G
$50.30
5 G
$201.00
25 G
$793.00

About This Item

Empirical Formula (Hill Notation):
C19H17N4S
CAS Number:
Molecular Weight:
333.43
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

$50.30


0 left in stock

Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥98% (TLC)
≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

15-25°C

SMILES string

S1CN([C@@H](C1)C(=O)O)C(=O)OCC2c3c(cccc3)c4c2cccc4

InChI

1S/C19H17NO4S/c21-18(22)17-10-25-11-20(17)19(23)24-9-16-14-7-3-1-5-12(14)13-6-2-4-8-15(13)16/h1-8,16-17H,9-11H2,(H,21,22)/t17-/m0/s1

InChI key

HUWNZLPLVVGCMO-KRWDZBQOSA-N

General description

A building block for the introduction of thiaproline (Thz) during Fmoc SPPS. Thz has been employed as a masked cysteine residue to prevent self-ligation during native chemical ligation reactions (NCL) of peptide thioesters bearing an N-terminal Cys residue [6]. Once ligation is complete, the N-terminal Cys residue of the resultant peptide is unmasked by ring-opening the Thz residue by treatment with methoxyamine, thereby enabling its subsequent ligation to another peptide thioester fragment.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] M. Villain, et al. in ′Peptides: The wave of the future, Proc. Second Int.& 17th American Peptide Symposium′, M. Lebl & R A. Houghten (Eds), American peptide Society, 2001, pp. 107.

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(011A)): ≥ 98 %
Assay (HPLC, area%): ≥ 99.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service