Skip to Content
Merck
All Photos(1)

Documents

8.52136

Sigma-Aldrich

Fmoc-Ser(Ac3AcNH-α-Gal)-OH

Novabiochem®

Synonym(s):

Fmoc-Ser(Ac3AcNH-α-Gal)-OH, N-α-Fmoc-O-(2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-galactopyranosyl)-L-serine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H36N2O13
CAS Number:
Molecular Weight:
656.63
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥95% (TLC)
≥95.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

15-25°C

InChI

1S/C32H36N2O13/c1-16(35)33-27-29(46-19(4)38)28(45-18(3)37)26(15-42-17(2)36)47-31(27)43-14-25(30(39)40)34-32(41)44-13-24-22-11-7-5-9-20(22)21-10-6-8-12-23(21)24/h5-12,24-29,31H,13-15H2,1-4H3,(H,33,35)(H,34,41)(H,39,40)/t25-,26+,27+,28-,29+,31-/m0/s1

InChI key

ORICVOOXZDVFIP-VOZJJELXSA-N

General description

The O-glycosidic linkage and the O-acetyl protection in this building block is stable to both piperidine and TFA [1], making it completely compatible with standard protocols in Fmoc solid phase peptide synthesis. Removal of the acetyl protecting groups is best carried out by treatment of the peptidyl resin with methanolic ammonia prior to cleavage.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Fmoc SPPS of Peptides containing Post-Translational Modifications


Literature references

[1] J. Kihlberg in ′Fmoc solid phase peptide synthesis: a practical approach′, W.C. Chan & P.D. White (Eds.), Oxford University Press, Oxford, 2000, pp. 195.

Linkage

Replaces: 04-12-8103

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(CMA2)): ≥ 95 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Identity (NMR, 1H): passes test
Identity (NMR): passes test
NMR-spectrum: passes test

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

We provide an overview of our available reagents, together with recommendations and details of their use for synthesis of peptides containing post-translationally modified amino acids.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service