Skip to Content
Merck
All Photos(1)

Key Documents

8.52095

Sigma-Aldrich

Fmoc-Lys(Mca)-OH

≥97.0% (HPLC), for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-Lys(Mca)-OH, N-α-Fmoc-N-ε-7-methoxycoumarin-4-acetyl-L-lysine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$644.00
1 G
$981.00
5 G
$4,000.00

$644.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$644.00
1 G
$981.00
5 G
$4,000.00

About This Item

Empirical Formula (Hill Notation):
C33H32N2O8
CAS Number:
Molecular Weight:
584.62
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

$644.00


Check Cart for Availability

Request a Bulk Order

Product Name

Fmoc-Lys(Mca)-OH, Novabiochem®

Quality Level

product line

Novabiochem®

Assay

≥97% (TLC)
≥97.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

−20°C

SMILES string

N([C@@H](CCCCNC(=O)Cc4c5c([o][c](c4)=O)cc(cc5)OC)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C33H32N2O8/c1-41-21-13-14-22-20(17-31(37)43-29(22)18-21)16-30(36)34-15-7-6-12-28(32(38)39)35-33(40)42-19-27-25-10-4-2-8-23(25)24-9-3-5-11-26(24)27/h2-5,8-11,13-14,17-18,27-28H,6-7,12,15-16,19H2,1H3,(H,34,36)(H,35,40)(H,38,39)/t28-/m0/s1

InChI key

BWJJONDPLYEAMC-NDEPHWFRSA-N

General description

A modified lysine derivative for the preparation of fluorogenically-labeled peptides by Fmoc SPPS [1]. The Mca group fluoresces at 405 nm when stimulated at 340 nm, and is most commonly used in conjunction with Dabcyl and 2,4-dinitrophenyl quenching groups. Recently, Fmoc-Lys(Mca)-OH has been used to prepare lanthanide-based luminescent proteins by native chemical ligation [2]. Lys(Mca) was found to be stable to HF cleavage conditions used to synthesize the thioester fragments needed for ligation.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Chromogenic Labeling of Synthetic Peptides

Literature references

[1] J. L. Lauer-Fields, et al. (2001) Biochemistry, 40, 5795.
[2] C. F. W. Becker, et al. (2004) Bioconjugate Chem., 15, 1118.

Linkage

Replaces: 04-12-1233

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.0 % (a/a)
Purity (TLC(011A)): ≥ 97 %
Assay (HPLC, area%): ≥ 97.0 % (a/a)
Water (K. F.): ≤ 2.0 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Chromogenic and fluorogenic derivatives are invaluable tools for biochemistry, having numerous applications in enzymology, protein chemistry, immunology and histochemistry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service