Skip to Content
Merck
All Photos(1)

Key Documents

8.52000

Sigma-Aldrich

Fmoc-Thr(tBu)-OH

Novabiochem®

Synonym(s):

Fmoc-Thr(tBu)-OH, N-α-Fmoc-O-t.-butyl-L-threonine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$112.00
100 G
$337.00
250 G
$770.00
500 G
$1,350.00
1 KG
$2,480.00
2 KG
$4,800.00
5 KG
$10,830.00

$112.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
$112.00
100 G
$337.00
250 G
$770.00
500 G
$1,350.00
1 KG
$2,480.00
2 KG
$4,800.00
5 KG
$10,830.00

About This Item

Empirical Formula (Hill Notation):
C23H27NO5
CAS Number:
Molecular Weight:
397.46
MDL number:
UNSPSC Code:
12352209
EC Index Number:
276-261-1
NACRES:
NA.22

$112.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥98% (TLC)
≥98.0% (acidimetric)
≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

125-135 °C

application(s)

peptide synthesis

functional group

hydroxyl

storage temp.

2-30°C

SMILES string

N([C@@H]([C@H](OC(C)(C)C)C)C(=O)[O-])C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C23H27NO5/c1-14(29-23(2,3)4)20(21(25)26)24-22(27)28-13-19-17-11-7-5-9-15(17)16-10-6-8-12-18(16)19/h5-12,14,19-20H,13H2,1-4H3,(H,24,27)(H,25,26)/p-1/t14-,20+/m1/s1

InChI key

LZOLWEQBVPVDPR-VLIAUNLRSA-M

General description

High purity Fmoc-protected amino acid for research and process production of peptides, with very low levels of dipeptide, free-amino acids and acetic acid impurities.
Standard building block for introduction of threonine amino-acid residues by Fmoc SPPS

Associated Protocols and Technical Articles
Fmoc-amino acids for Peptide Production
Cleavage and Deprotection Protocols for Fmoc SPPS

Application


  • Protocol for Facile Synthesis of Fmoc-N-Me-AA-OH Using 2-CTC Resin as Temporary and Reusable Protecting Group: This study demonstrates the effective use of Fmoc-Thr(tBu)-OH in the synthesis of N-methyl amino acids, which are crucial in developing peptidomimetic drugs (Román et al., 2023).

  • Controlled Morphological Changes in Self-Assembled Structures Formed by Fmoc Variants of Threonine and Serine: Explores how Fmoc-Thr(tBu)-OH influences the self-assembly and structural morphology of peptidic materials, relevant in material science (Kshtriya et al., 2021).

  • Solid-phase Synthesis of C-terminus Cysteine Peptide Acids: This article includes the use of Fmoc-Thr(tBu)-OH in the synthesis of peptides containing cysteine, which are significant in both drug design and biochemical studies (Mthembu et al., 2022).

  • Polymer-Supported Stereoselective Synthesis of Benzoxazinothiadiazepinone 6,6-dioxides: Discusses the use of Fmoc-Thr(tBu)-OH in the stereoselective synthesis of novel organic compounds with potential applications in medicinal chemistry (Králová et al., 2017).


Linkage

Replaces: 04-12-1000

Analysis Note

Colour (visual): white to off white
Appearance of substance (visual): powder
Colour index (0,5 M in DMF): ≤ 150 Hazen
Identity (IR): passes test
Enantiomeric purity: ≥ 99.7 % (a/a)
Purity (HPLC): ≥ 99.0 % (a/a)
Fmoc-ß-Ala-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-ß-Ala-Thr(tBu)-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Thr(tBu)-Thr(tBu)-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Thr-OH (HPLC): ≤ 0.1 % (a/a)
Assay free amino acid (GC): ≤ 0.2 %
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (25 mmole in 50 ml DMF): clearly soluble
Assay (acidimetric): ≥ 98.0 %
Water (K. F.): ≤ 2.0 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.02 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service