Skip to Content
Merck
All Photos(1)

Key Documents

8.51210

Sigma-Aldrich

1-Acetylimidazole

Novabiochem®

Synonym(s):

1-Acetylimidazole

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$78.20
100 G
$234.00
250 G
$398.00
500 G
$714.00

$78.20


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
$78.20
100 G
$234.00
250 G
$398.00
500 G
$714.00

About This Item

Empirical Formula (Hill Notation):
C5H6N2O
CAS Number:
Molecular Weight:
110.11
MDL number:
UNSPSC Code:
12352100
EC Index Number:
219-577-7
NACRES:
NA.22

$78.20


Estimated to ship on22 May 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

form

powder

manufacturer/tradename

Novabiochem®

mp

99-105 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

[n]1(cncc1)C(=O)C

InChI

1S/C5H6N2O/c1-5(8)7-3-2-6-4-7/h2-4H,1H3

InChI key

VIHYIVKEECZGOU-UHFFFAOYSA-N

General description

Reagent for acetylation of amines under mild conditions.

Analysis Note

Appearance of substance (visual): powder
Colour (visual): white to off white
Assay (HCIO4): ≥ 97.5 %
Assay (GG, area%): ≥ 97.5 % (a/a)
Melting range (lower value): ≥ 90 °C
Melting range (upper value): ≤ 105 °C
Solubility (1 mmole in 2 ml DMF): clearly soluble
Identity (IR): passes test

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service