Skip to Content
Merck
All Photos(1)

Key Documents

8.51097

Sigma-Aldrich

6-Azido-hexanoic acid

Novabiochem®

Synonym(s):

6-Azido-hexanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$312.00
5 G
$1,560.00

$312.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
1 G
$312.00
5 G
$1,560.00

About This Item

Empirical Formula (Hill Notation):
C6H11N3O2
CAS Number:
Molecular Weight:
157.17
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

$312.00


In StockDetails


Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥98.0% (GC)

form

oil

reaction suitability

reaction type: click chemistry

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

15-25°C

SMILES string

[N+](=[N-])=NCCCCCC(=O)O

InChI

1S/C6H11N3O2/c7-9-8-5-3-1-2-4-6(10)11/h1-5H2,(H,10,11)

InChI key

JCORXJUUSVCJEP-UHFFFAOYSA-N

General description

6-Azido-hexanoic acid is a building block used to introduce a clickable azido group to amino group for use in click chemistry, protein labeling, peptide synthesis, and surface modifications.

Application

Recent applications of 6-azido-hexanoic acid involving click chemistry:
  • In the synthesis of cellulose-based click scaffolds.[1]
  • In the functionalization of a novel ethynylzinc (II) phthalocyanine compound via click chemistry.[2]
  • In the production of peptide microarrays via catalyst-free click chemistry.[3]

Analysis Note

Colour (visual): colorless to slight yellow to yellow
Appearance of substance (visual): oil
Identity (IR): passes test
Assay (GC, area%): ≥ 98.0 % (a/a)

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ADIBO-based ?click? chemistry for diagnostic peptide micro-array fabrication: physicochemical and assay characteristics
Prim D, et al.
Molecules (Basel), 18, 9833-9833 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service