Skip to Content
Merck
All Photos(1)

Key Documents

8.51032

Sigma-Aldrich

Trt-NH-(PEG)₂-NH₂ (15 atoms)

Novabiochem®

Synonym(s):

Trt-NH-(PEG)₂-NH₂ (15 atoms), O-(N-Trt-3-aminopropyl)-O′-(3-aminpropyl)-diethyleneglycol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$269.00

$269.00


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
$269.00

About This Item

Empirical Formula (Hill Notation):
C29H38N2O3
CAS Number:
Molecular Weight:
462.62
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

$269.00


Estimated to ship on14 April 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥97% (TLC)

form

oil

reaction suitability

reaction type: Pegylations

manufacturer/tradename

Novabiochem®

transition temp

flash point 110 °C (External MSDS)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

N(CCCOCCOCCOCCCN)C(c3ccccc3)(c2ccccc2)c1ccccc1

InChI

1S/C29H38N2O3/c30-18-10-20-32-22-24-34-25-23-33-21-11-19-31-29(26-12-4-1-5-13-26,27-14-6-2-7-15-27)28-16-8-3-9-17-28/h1-9,12-17,31H,10-11,18-25,30H2

InChI key

FGUBGJQGKWQTBO-UHFFFAOYSA-N

General description

This mono-protected diamino functionalized PEG is a useful building block for the introduction of a PEG spacer into molecules containing a carboxylic acid functionality. Removal of the Trt group can be effected using 1-5% TFA in combination with 2-3% TES in DCM.

Linkage

Replaces: 01-63-0143

Analysis Note

Color (visual): pale yellow to yellow to beige
Appearance of substance (visual): oil
Identity (IR): passes test
Purity (TLC(CMA1)): ≥ 97 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - (External MSDS)

Flash Point(C)

110 °C - (External MSDS)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service