Skip to Content
Merck
All Photos(1)

Key Documents

8.51017

Sigma-Aldrich

Boc-amino-oxyacetic acid

Novabiochem®

Synonym(s):

Boc-amino-oxyacetic acid, Boc-NH-O-CH 2COOH, Boc-Aoa-OH

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$116.00
5 G
$470.00

$116.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
$116.00
5 G
$470.00

About This Item

Empirical Formula (Hill Notation):
C7H13NO5
CAS Number:
Molecular Weight:
191.18
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

$116.00


Estimated to ship on27 May 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥98% (TLC)

form

powder

reaction suitability

reagent type: ligand

manufacturer/tradename

Novabiochem®

mp

115 °C (decomposes)

application(s)

peptide synthesis

storage temp.

2-30°C

SMILES string

N(OCC(=O)O)C(=O)OC(C)(C)C

InChI

1S/C7H13NO5/c1-7(2,3)13-6(11)8-12-4-5(9)10/h4H2,1-3H3,(H,8,11)(H,9,10)

InChI key

QBXODCKYUZNZCY-UHFFFAOYSA-N

General description

This reagent can be used to introduce a hydroxylamine functionality to N-terminal or side-chain amino groups. The use of this reagent may lead to double acylation of the hydroxylamine nitrogen. Hydroxylamine-labeled peptides prepared in this manner can be ligated in aqueous solution at pH 3.5 to aldehyde-containing peptides via oxime formation.

Linkage

Replaces: 01-63-0060

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(CMA2)): ≥ 98 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service