Skip to Content
Merck
All Photos(1)

Key Documents

8.51015

Sigma-Aldrich

Dde-OH

Novabiochem®

Synonym(s):

Dde-OH, 2-Acetyldimedone

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$289.00
25 G
$1,160.00
100 G
$3,520.00

$289.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
$289.00
25 G
$1,160.00
100 G
$3,520.00

About This Item

Empirical Formula (Hill Notation):
C10H14O3
CAS Number:
Molecular Weight:
182.22
MDL number:
UNSPSC Code:
12352115
NACRES:
NA.22

$289.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥98% (TLC)

form

powder

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

InChI

1S/C10H14O3/c1-6(11)9-7(12)4-10(2,3)5-8(9)13/h12H,4-5H2,1-3H3

InChI key

DIJDAMFZOHSRID-UHFFFAOYSA-N

General description

Reagent for the introduction of the Dde protecting group [1,2,3,4].

Associated Protocols and Technical Articles
Guide to Selection of Orthogonally -Protected Building Blocks for Fmoc SPPS

Literature references

[1] B. W. Bycroft, et al. (1993) J. Chem. Soc., Chem. Commun., 778.
[2] I. A. Nash, et al. (1996) Tetrahedron Lett., 37, 2625.
[3] F. Berst, et al. (2000) Tetrahedron Lett., 41, 6649.
[4] A. N. Acharaya, et al. (2001) J. Comb. Chem., 3, 612.

Linkage

Replaces: 01-63-0033

Analysis Note

Color (visual): white to slight yellow to yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (DC(012A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service