Skip to Content
Merck
All Photos(1)

Documents

8.51000

Sigma-Aldrich

HMPA

4-Hydroxymethylphenoxyacetic acid Novabiochem®

Synonym(s):

HMPA, 4-Hydroxymethylphenoxyacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O4
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥98.0% (TLC)
≥99.0% (HPLC)

form

powder

reaction suitability

reagent type: linker

manufacturer/tradename

Novabiochem®

mp

110-120 °C

application(s)

peptide synthesis

storage temp.

2-8°C

InChI

1S/C9H10O4/c10-5-7-1-3-8(4-2-7)13-6-9(11)12/h1-4,10H,5-6H2,(H,11,12)/p-1

InChI key

VUCNQOPCYRJCGQ-UHFFFAOYSA-M

General description

Standard TFA-labile linker for the preparation of peptide acids by Fmoc SPPS.

Linkage

Replaces: 01-60-0017

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(0811)): ≥ 98.0 %
Assay (HPLC, area%): ≥ 99.0 % (a/a)
Water (K. F.): ≤ 1.00 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service