Skip to Content
Merck
All Photos(1)

Documents

8.45045

Sigma-Aldrich

Iodomethane

(stabilised with silver) Msynthplus

Synonym(s):

Iodomethane, Methyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
141.94
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-819-5

vapor pressure

540 hPa ( 25 °C)

Assay

≥99% (GC)

form

liquid

potency

79.84 mg/kg LD50, oral (Rat)

expl. lim.

8.5-66 % (v/v)

pH

5.2 (25 °C in H2O)

bp

42 °C/1013 hPa

mp

-66 °C

solubility

8.66 g/L

density

2.28 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/CH3I/c1-2/h1H3

InChI key

INQOMBQAUSQDDS-UHFFFAOYSA-N

General description

(stabilised with silver) Msynthplus

Application

Iodomethane (stabilised with silver) Msynthplus. CAS 74-88-4, pH 5.2 (H₂O, 25 °C).

Analysis Note

Color (visual)colourless to yellowAppearance of substance (visual)clearAssay (GC, area%)≥ 99.0 % (a/a)Identity (IR)passes testThe analysis of residual solvents is not possible due to the physicochemical properties of the material.

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(C)

32.1 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service