Skip to Content
Merck
All Photos(1)

Key Documents

8.44011

Sigma-Aldrich

Ethylene carbonate

for synthesis

Synonym(s):

Ethylene carbonate, 1,3-Dioxolane-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$63.20

$63.20


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
1 KG
$63.20

About This Item

CAS Number:
Molecular Weight:
88.06
MDL number:
UNSPSC Code:
12352115
EC Index Number:
202-510-0
NACRES:
NA.22

$63.20


Estimated to ship on30 May 2025


Request a Bulk Order

vapor pressure

0.21 hPa ( 20 °C)

Quality Level

form

crystals

autoignition temp.

465 °C

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

3.6-16.1 % (v/v)

pH

7 (20 °C, 200 g/L in H2O)

bp

248 °C/1013 hPa

mp

36 °C

transition temp

flash point 143 °C

solubility

214 g/L

density

1.32 g/cm3 at 40 °C

storage temp.

2-30°C

SMILES string

O1CCOC1=O

InChI

1S/C3H4O3/c4-3-5-1-2-6-3/h1-2H2

InChI key

KMTRUDSVKNLOMY-UHFFFAOYSA-N

General description

for synthesis

Application

Ethylene carbonate for synthesis. CAS 96-49-1, pH 7 (200 g/l, H₂O, 20 °C).

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

289.4 °F - closed cup

Flash Point(C)

143 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service