Skip to Content
Merck
All Photos(1)

Key Documents

8.43742

Sigma-Aldrich

Titanium(IV) bis(acetylacetonate) diisopropoxide

(75% solution in 2-propanol) for synthesis

Synonym(s):

Diisopropoxytitanium bis(acetylacetonate) solution, Ti(acac)2OiPr2

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$71.30
500 ML
$254.00

$71.30


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$71.30
500 ML
$254.00

About This Item

UNSPSC Code:
12352103
NACRES:
NA.22

$71.30


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

form

liquid

transition temp

flash point 12 °C

density

1.01 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Assay (ex Ti): 72.0 - 78.0 %
Density (d 20 °C/ 4 °C): 0.990 - 1.020
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service