Skip to Content
Merck
All Photos(1)

Key Documents

8.43658

Sigma-Aldrich

Trimethoxyphenylsilane

for synthesis

Synonym(s):

Trimethoxyphenylsilane

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$143.00

$143.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
50 ML
$143.00

About This Item

Linear Formula:
(CH3O)3SiC6H5
CAS Number:
Molecular Weight:
198.29
MDL number:
UNSPSC Code:
12352103
EC Index Number:
221-066-9
NACRES:
NA.22

$143.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥96.5% (GC)

form

liquid

transition temp

flash point 83 °C

density

1.064 g/cm3

storage temp.

2-30°C

SMILES string

[Si](OC)(OC)(OC)c1ccccc1

InChI

1S/C9H14O3Si/c1-10-13(11-2,12-3)9-7-5-4-6-8-9/h4-8H,1-3H3

InChI key

ZNOCGWVLWPVKAO-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.5 % (a/a)
Density (d 20 °C/ 4 °C): 1.062 - 1.066
Identity (IR): passes test

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - STOT RE 2 Oral

Target Organs

Bladder

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service