Skip to Content
Merck
All Photos(1)

Key Documents

8.43602

Sigma-Aldrich

N-Ethyl-o-toluidine

for synthesis

Synonym(s):

N-Ethyl-o-toluidine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$126.00

$126.00


Estimated to ship on02 May 2025



Select a Size

Change View
100 ML
$126.00

About This Item

Empirical Formula (Hill Notation):
C9H13N
CAS Number:
Molecular Weight:
135.21
MDL number:
UNSPSC Code:
12352116
EC Index Number:
202-354-3
NACRES:
NA.22

$126.00


Estimated to ship on02 May 2025


vapor pressure

1 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

autoignition temp.

445 °C

potency

2400 mg/kg LD50, oral (Rat)

expl. lim.

1.2-5.9 % (v/v)

bp

217 °C/1013 hPa

mp

<-15 °C

transition temp

flash point 88 °C

solubility

1 g/L

density

0.95 g/mL

storage temp.

2-30°C

SMILES string

N(CC)c1c(cccc1)C

InChI

1S/C9H13N/c1-3-10-9-7-5-4-6-8(9)2/h4-7,10H,3H2,1-2H3

InChI key

MWOUGPLLVVEUMM-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.947 - 0.951
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service