Skip to Content
Merck
All Photos(1)

Key Documents

8.43070

Sigma-Aldrich

Isobutyl stearate

for synthesis

Synonym(s):

Isobutyl stearate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$36.70
1 L
$64.60
2.5 L
$133.00
8.5 KG
$681.00

$36.70


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$36.70
1 L
$64.60
2.5 L
$133.00
8.5 KG
$681.00

About This Item

Empirical Formula (Hill Notation):
C22H44O2
CAS Number:
Molecular Weight:
340.58
MDL number:
UNSPSC Code:
12352108
EC Index Number:
211-466-1
NACRES:
NA.22
grade:
for synthesis
synthesis grade

$36.70


Estimated to ship on31 May 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

form

solid

dilution

(for synthesis)

mp

28.9 °C

density

0.85 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC(C)C)C(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C22H44O2/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-22(23)24-20-21(2)3/h21H,4-20H2,1-3H3

InChI key

ORFWYUFLWUWSFM-UHFFFAOYSA-N

Analysis Note

Saponification value: 170 - 179
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service