Skip to Content
Merck
All Photos(1)

Key Documents

8.41638

Sigma-Aldrich

Trimethyloxonium tetrafluoroborate

for synthesis

Synonym(s):

Trimethyloxonium tetrafluoroborate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$182.00

$182.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 G
$182.00

About This Item

Empirical Formula (Hill Notation):
C3H9BF4O
CAS Number:
Molecular Weight:
147.91
MDL number:
UNSPSC Code:
12352107
EC Index Number:
206-994-4
NACRES:
NA.22

$182.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥95.0% (acidimetric)

form

crystals

mp

202-203 °C (sublimed)

storage temp.

2-8°C

SMILES string

[B+3].[F-].[F-].[F-].[F-].[O+](C)(C)C

InChI

1S/C3H9O.B.4FH/c1-4(2)3;;;;;/h1-3H3;;4*1H/q+1;+3;;;;/p-4

InChI key

KAAQTDVQNWMZJT-UHFFFAOYSA-J

Analysis Note

Assay (acidimetric): ≥ 95.0 %
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service