Skip to Content
Merck
All Photos(1)

Key Documents

8.41620

Sigma-Aldrich

Cyclohexylmethanol

for synthesis

Synonym(s):

Cyclohexylmethanol

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$142.00
250 ML
$406.00

$142.00


Estimated to ship on31 May 2025



Select a Size

Change View
50 ML
$142.00
250 ML
$406.00

About This Item

Empirical Formula (Hill Notation):
C7H14O
CAS Number:
Molecular Weight:
114.19
MDL number:
UNSPSC Code:
12352104
EC Index Number:
202-857-8
NACRES:
NA.22
grade:
for synthesis
synthesis grade

$142.00


Estimated to ship on31 May 2025


grade

for synthesis
synthesis grade

Quality Level

Assay

≥99.0% (GC)

autoignition temp.

330 °C

dilution

(for synthesis)

transition temp

flash point 77 °C

density

0.93 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OCC1CCCCC1

InChI

1S/C7H14O/c8-6-7-4-2-1-3-5-7/h7-8H,1-6H2

InChI key

VSSAZBXXNIABDN-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.927 - 0.934
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service