Skip to Content
Merck
All Photos(1)

Key Documents

8.41554

Sigma-Aldrich

Benzotrifluoride

for synthesis

Synonym(s):

Benzotrifluoride

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$84.20

$84.20


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$84.20

About This Item

Empirical Formula (Hill Notation):
C7H5F3
CAS Number:
Molecular Weight:
146.11
MDL number:
UNSPSC Code:
12352101
EC Index Number:
202-635-0
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
100-103 °C/1013 hPa
vapor pressure:
53 hPa ( 25 °C)

$84.20


Estimated to ship on28 May 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

53 hPa ( 25 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

650 °C

potency

15000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

expl. lim.

1.4-9.3 % (v/v)

dilution

(for synthesis)

bp

100-103 °C/1013 hPa

mp

-29 °C

transition temp

flash point 12 °C

solubility

0.45 g/L (Hydrolysis)

density

1.197 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

FC(F)(F)c1ccccc1

InChI

1S/C7H5F3/c8-7(9,10)6-4-2-1-3-5-6/h1-5H

InChI key

GETTZEONDQJALK-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.186 - 1.190
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 2 - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service