Skip to Content
Merck
All Photos(1)

Key Documents

8.41133

Sigma-Aldrich

3,4-Dichlorobenzyl alcohol

for synthesis

Synonym(s):

3,4-Dichlorobenzyl alcohol, 3,4-Dichloro-1-(hydroxymethyl)-benzene

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$90.00

$90.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
10 G
$90.00

About This Item

Empirical Formula (Hill Notation):
C7H6Cl2O
CAS Number:
Molecular Weight:
177.03
MDL number:
UNSPSC Code:
12352104
EC Index Number:
217-299-0
NACRES:
NA.22

$90.00


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

solid

bp

148-151 °C

mp

38-39 °C

transition temp

flash point >110 °C

storage temp.

2-30°C

SMILES string

Clc1c(ccc(c1)CO)Cl

InChI

1S/C7H6Cl2O/c8-6-2-1-5(4-10)3-7(6)9/h1-3,10H,4H2

InChI key

FVJIUQSKXOYFKG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 36 °C
Melting range (upper value): ≤ 39 °C
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service