Skip to Content
Merck
All Photos(1)

Key Documents

8.41086

Sigma-Aldrich

Ethyl 4-dimethylaminobenzoate

for synthesis

Synonym(s):

Ethyl 4-dimethylaminobenzoate, 4-Dimethylaminobenzoic acid ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$172.00

$172.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 G
$172.00

About This Item

Empirical Formula (Hill Notation):
C11H15NO2
CAS Number:
Molecular Weight:
193.24
MDL number:
UNSPSC Code:
12352108
EC Index Number:
233-634-3
NACRES:
NA.22

$172.00


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

Assay

≥99.0% (GC)

form

powder

mp

63-65 °C

storage temp.

2-30°C

SMILES string

N(C)(C)c1ccc(cc1)C(=O)OCC

InChI

1S/C11H15NO2/c1-4-14-11(13)9-5-7-10(8-6-9)12(2)3/h5-8H,4H2,1-3H3

InChI key

FZUGPQWGEGAKET-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 63 °C
Melting range (upper value): ≤ 65 °C
Identity (IR): passes test

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service