Skip to Content
Merck
All Photos(1)

Key Documents

8.41018

Sigma-Aldrich

Ethyl salicylate

for synthesis

Synonym(s):

Ethyl salicylate, Salicylic acid ethyl ester, Ethyl-2-hydroxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O3
CAS Number:
Molecular Weight:
166.17
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-265-5
form:
liquid
Assay:
≥98.0% (GC)
Pricing and availability is not currently available.

vapor pressure

0.1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

1320 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.1 % (v/v)

bp

232 °C/1013 hPa

mp

1.3 °C

transition temp

flash point 100 °C

solubility

0.25 g/L

density

1.13 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C9H10O3/c1-2-12-9(11)7-5-3-4-6-8(7)10/h3-6,10H,2H2,1H3

InChI key

GYCKQBWUSACYIF-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.128 - 1.132
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

212.0 °F

Flash Point(C)

100 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service