Skip to Content
Merck
All Photos(1)

Key Documents

8.40076

Sigma-Aldrich

Pentaethylenehexamine

(mixture of isomers) for synthesis

Synonym(s):

Pentaethylenehexamine, 3,6,9,12-Tetraazatetradecane-1,14-diamine

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$53.50
1 L
$142.00

$53.50


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
25 ML
$53.50
1 L
$142.00

About This Item

Empirical Formula (Hill Notation):
C10H28N6
CAS Number:
Molecular Weight:
232.37
MDL number:
UNSPSC Code:
12352116
EC Index Number:
223-775-9
NACRES:
NA.22

$53.50


Estimated to ship on30 May 2025


Request a Bulk Order

vapor pressure

<1 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

360 °C

pH

11 (10 g/L in H2O)

mp

-35 °C

transition temp

flash point 175 °C

density

1.003 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CCNCCNCCN)CCNCCN

InChI

1S/C10H28N6/c11-1-3-13-5-7-15-9-10-16-8-6-14-4-2-12/h13-16H,1-12H2

InChI key

LSHROXHEILXKHM-UHFFFAOYSA-N

Analysis Note

Assay of N: 33.0 - 35.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service