8.24449
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid
for synthesis
Synonym(s):
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid, (1R,2R)-(-)-Hexahydrophthalic acid
Select a Size
$33.20
Estimated to ship on30 May 2025
Select a Size
About This Item
$33.20
Estimated to ship on30 May 2025
Recommended Products
Quality Level
Assay
≥99.0% (acidimetric)
mp
177-180 °C
storage temp.
2-30°C
SMILES string
OC(=O)[C@H]1[C@@H](CCCC1)C(=O)O
InChI
1S/C8H12O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h5-6H,1-4H2,(H,9,10)(H,11,12)/t5-,6-/m1/s1
InChI key
QSAWQNUELGIYBC-PHDIDXHHSA-N
Analysis Note
Melting range (lower value): ≥ 177 °C
Melting range (upper value): ≤ 180 °C
Spec. rotation [α²0/D (c=1 in Aceton): -20.0 - -19.0 °
Identity (IR): passes test
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service