Skip to Content
Merck
All Photos(1)

Key Documents

8.22321

Sigma-Aldrich

Phosphorus tribromide

for synthesis

Synonym(s):

Phosphorus tribromide, Phosphorus(III) bromide

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$87.80
250 ML
$401.00
6 KG
$1,740.00

$87.80


Available to ship on01 April 2025Details


Request a Bulk Order

Select a Size

Change View
50 ML
$87.80
250 ML
$401.00
6 KG
$1,740.00

About This Item

Empirical Formula (Hill Notation):
Br3P
CAS Number:
Molecular Weight:
270.69
MDL number:
UNSPSC Code:
12352307
EC Index Number:
232-178-2
NACRES:
NA.22

$87.80


Available to ship on01 April 2025Details


Request a Bulk Order

vapor pressure

2.9 hPa ( 20 °C)

Quality Level

form

liquid

refractive index

n20/589 1.6945

bp

173-175 °C/1013 hPa

mp

-40 °C

density

2.85 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

P(Br)(Br)Br

InChI

1S/Br3P/c1-4(2)3

InChI key

IPNPIHIZVLFAFP-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 98.0 %
Appearance of substance (visual): clear to light turbid
Density (d 20 °C/ 4 °C): 2.870 - 2.890

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service