Skip to Content
Merck
All Photos(1)

Key Documents

8.22302

Sigma-Aldrich

Pyrogallol

for synthesis

Synonym(s):

Pyrogallol, Pyrogallic acid, 1,2,3-Trihydroxybenzene

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$72.90
250 G
$285.00
1 KG
$586.00

$72.90


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
$72.90
250 G
$285.00
1 KG
$586.00

About This Item

Linear Formula:
C6H3(OH)3
CAS Number:
Molecular Weight:
126.11
MDL number:
UNSPSC Code:
12352104
EC Index Number:
201-762-9
NACRES:
NA.22
grade:
for synthesis
bp:
309 °C/1013 hPa
vapor pressure:
2-4 hPa ( 140 °C)

$72.90


Estimated to ship on22 May 2025


Request a Bulk Order

grade

for synthesis

Quality Level

vapor pressure

2-4 hPa ( 140 °C)

Assay

≥99% (acidimetric)

form

solid

potency

790 mg/kg LD50, oral (Rat)

dilution

(for synthesis)

pH

4-5 (20 °C, 50 g/L in H2O)

bp

309 °C/1013 hPa

mp

131-135 °C

solubility

400 g/L

density

1.45 g/cm3 at 20 °C

bulk density

600 kg/m3

storage temp.

2-30°C

SMILES string

Oc1c(cccc1O)O

InChI

1S/C6H6O3/c7-4-2-1-3-5(8)6(4)9/h1-3,7-9H

InChI key

WQGWDDDVZFFDIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Antioxidant and therapeutic potential: Pyrogallol demonstrates significant anti-inflammatory, sedative, antidiabetic, and antioxidant capabilities across both in vitro and in vivo models, as evidenced by a comprehensive phenolic profiling of Atractylis aristata Batt (Abid et al., 2024).

  • Photo-switchable collector applications: The utility of pyrogallol-derived compounds as photo-switchable collectors in the flotation process for lithium aluminate recycling has been explored, highlighting its role in enhancing raw material recovery (Zgheib et al., 2024).

  • Biomedical applications in hydrogels: Pyrogallol is integrated into the design of a hydrogel, focusing on biomedical applications such as wound healing and drug delivery, offering insights into the versatile uses of pyrogallol in medical materials (Weian et al., 2024).

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Melting range (lower value): ≥ 132 °C
Melting range (upper value): ≤ 135 °C
Identity (IR): passes test

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service