Skip to Content
Merck
All Photos(3)

Key Documents

8.22277

Supelco

Ethyl acetate

EMPLURA®

Synonym(s):

Ethyl ethanoate, Acetic ester, Ethyl ester, EtOAc

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 L
$53.50
5 L
$96.30

$53.50


Estimated to ship on28 March 2025


Request a Bulk Order

Select a Size

Change View
2.5 L
$53.50
5 L
$96.30

About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
MDL number:
UNSPSC Code:
12352108
EC Index Number:
205-500-4
NACRES:
NA.05
grade:
for analytical purposes
for cleaning
for extraction
for preparative purposes
for synthesis
Assay:
≥99.5% (GC)
application(s):
environmental
food and beverages
industrial qc
pharmaceutical
bp:
76.5-77.5 °C (lit.)
vapor pressure:
73 mmHg ( 20 °C)

$53.50


Estimated to ship on28 March 2025


Request a Bulk Order

grade

for analytical purposes
for cleaning
for extraction
for preparative purposes
for synthesis

Quality Level

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

product line

EMPLURA®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

801 °F

potency

5620 mg/kg LD50, oral (Rat)
>18000 mg/kg LD50, skin (Rabbit)

expl. lim.

2.2-11.5 %, 38 °F

impurities

≤0.1% Water

evapn. residue

≤0.003%

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

transition temp

flash point -4 °C

solubility

85.3 g/L

density

0.902 g/mL at 25 °C (lit.)

application(s)

environmental
food and beverages
industrial qc
pharmaceutical

format

neat

storage temp.

2-30°C

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application


  • [Determination of 12 prohibited veterinary drug residues in pig urine by ultra high performance liquid chromatography-tandem mass spectrometry]: Ethyl acetate is utilized for sample preparation in this advanced method to detect drug residues in animal samples, emphasizing its role in veterinary pharmaceutical compliance (Wan et al., 2024).

  • Therapeutic effect of Ginkgetin on smoke-induced airway inflammation by down-regulating the c/EBPβ signaling pathway and CCL2 expression: This study explores the anti-inflammatory properties of Ginkgetin, where ethyl acetate is used in the extraction process, highlighting its utility in biomedical research (Tao et al., 2024).

  • Effect of methanol extract of Plectranthus esculentus N.E.Br tuber and its fractions on indices of benign prostatic hyperplasia in Wistar rats: Ethyl acetate is employed in the extraction of bioactive compounds for pharmacological studies, showing its critical role in therapeutic applications (Kanu et al., 2024).

  • Phytochemical evaluation of Ziziphus mucronata and Xysmalobium undulutum towards the discovery and development of anti-malarial drugs: Demonstrates the use of ethyl acetate in phytochemical screenings, contributing to the development of new anti-malarial medications (Buthelezi et al., 2024).

  • The First Records of the In Silico Antiviral and Antibacterial Actions of Molecules Detected in Extracts of Algerian Fir (Abies numidica De Lannoy) Using LC-MS/MS Analysis: This research utilizes ethyl acetate for extracting compounds from plants for antiviral and antibacterial testing, underscoring its importance in the study of natural products (Benouchenne et al., 2024).

Analysis Note

Purity (GC): ≥ 99.5 %
Identity (IR): conforms
Density (d 20 °C/ 4 °C): 0.899 - 0.901
Water: ≤ 0.1 %
Evaporation residue: ≤ 0.003 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F

Flash Point(C)

-4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service