Skip to Content
Merck
All Photos(2)

Key Documents

8.22194

Sigma-Aldrich

(-)-Quinine hydrochloride dihydrate

for synthesis

Synonym(s):

(-)-Quinine hydrochloride dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$81.10
100 G
$213.00

$81.10


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
$81.10
100 G
$213.00

About This Item

Empirical Formula (Hill Notation):
C20H25ClN2O2 · 2H2O
CAS Number:
Molecular Weight:
396.91
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-001-1
NACRES:
NA.22

$81.10


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

Assay

≥99.0% (acidimetric)

form

powder

potency

620 mg/kg LD50, oral (Rat)

pH

6.0 (20 °C, 10 g/L in H2O)

mp

115 °C (decomposition)

transition temp

flash point 122 °C

solubility

62.5 g/L

bulk density

310 kg/m3

storage temp.

2-30°C

SMILES string

Cl.N21[C@@H](C[C@H]([C@H](C2)C=C)CC1)[C@H](O)c3c4c(ncc3)ccc(c4)OC.O.O

InChI

1S/C20H24N2O2.ClH.2H2O/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;;;/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;1H;2*1H2/t13-,14+,19-,20+;;;/m0.../s1

InChI key

MPQKYZPYCSTMEI-FXXIJDEQSA-N

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Spec. rotation [α²0/D (calculated on dried substance): -258.0 - -245.0 °
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service