Skip to Content
Merck
All Photos(1)

Key Documents

8.22161

Sigma-Aldrich

4-Hydroxy-3,5-dimethoxybenzoic acid

for synthesis

Synonym(s):

4-Hydroxy-3,5-dimethoxybenzoic acid, 3,5-Dimethoxy-4-hydroxybenzoic acid, Syringic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$95.40

$95.40


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 G
$95.40

About This Item

Linear Formula:
3,5-(CH3O)2-4-(OH)C6H2COOH
CAS Number:
Molecular Weight:
198.17
MDL number:
UNSPSC Code:
12352106
EC Index Number:
208-486-8
NACRES:
NA.22

$95.40


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥96.0% (acidimetric)

form

powder

mp

204-207 °C

storage temp.

2-30°C

SMILES string

O(C)c1c(c(cc(c1)C(=O)O)OC)O

InChI

1S/C9H10O5/c1-13-6-3-5(9(11)12)4-7(14-2)8(6)10/h3-4,10H,1-2H3,(H,11,12)

InChI key

JMSVCTWVEWCHDZ-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 96.0 %
Melting range (lower value): ≥ 205 °C
Melting range (upper value): ≤ 209 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service