Skip to Content
Merck
All Photos(1)

Key Documents

8.22085

Sigma-Aldrich

di-Lithium oxalate

for synthesis

Synonym(s):

di-Lithium oxalate, Oxalic acid dilithium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$156.00

$156.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
$156.00

About This Item

Empirical Formula (Hill Notation):
C2Li2O4
CAS Number:
Molecular Weight:
101.90
MDL number:
UNSPSC Code:
12352107
EC Index Number:
209-054-1
NACRES:
NA.22

$156.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

form

crystals

solubility

66 g/L

storage temp.

2-30°C

SMILES string

[Li+].[Li+].[O-]C(=O)C(=O)[O-]

InChI

1S/C2H2O4.2Li/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;/q;2*+1/p-2

InChI key

YNQRWVCLAIUHHI-UHFFFAOYSA-L

Analysis Note

Assay (manganometric): ≥ 98.0 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service