Skip to Content
Merck
All Photos(1)

Key Documents

8.21943

Sigma-Aldrich

2-Ethylhexyl phosphate

(mixture of esters) for synthesis

Synonym(s):

2-Ethylhexyl phosphate, Phosphoric acid 2-ethylhexyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$51.80
1 L
$107.00

$51.80


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$51.80
1 L
$107.00

About This Item

Empirical Formula (Hill Notation):
C8H18O · xH3PO4
CAS Number:
Molecular Weight:
130.23
MDL number:
UNSPSC Code:
12352106
EC Index Number:
235-741-0
NACRES:
NA.22

$51.80


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

form

liquid

potency

3000 mg/kg LD50, oral (Rat)

pH

2.3 (20 °C, 10 g/L in H2O, as an emulsion)

bp

>180 °C/1013 hPa (decomposes)

transition temp

flash point >100 °C

density

1.02 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[P](=O)(O)(O)O.OCC(CCCC)CC

InChI

1S/C8H18O.H3O4P/c1-3-5-6-8(4-2)7-9;1-5(2,3)4/h8-9H,3-7H2,1-2H3;(H3,1,2,3,4)

InChI key

FZQYZMODYSBPES-UHFFFAOYSA-N

Related Categories

Analysis Note

Colour (visual): colourless to brown
Appearance (visual): clear
Assay (Mono- and Diester) (acidimetric): ≥ 95.0 %
Assay of monoester (acidimetric): 30.0 - 50.0 %
Assay of diester (acidimetric): 40.0 - 70.0 %
Density (d 20 °C/ 4 °C): 1.015 - 1.025
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

212.0 °F

Flash Point(C)

> 100 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service