Skip to Content
Merck
All Photos(2)

Key Documents

8.21897

Sigma-Aldrich

N,N,N′,N′-Tetramethylguanidine

for synthesis

Synonym(s):

N,N,N′,N′-Tetramethylguanidine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$94.40
250 ML
$143.00

$94.40


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$94.40
250 ML
$143.00

About This Item

Linear Formula:
[(CH3)2N]2C=NH
CAS Number:
Molecular Weight:
115.18
MDL number:
UNSPSC Code:
12352129
EC Index Number:
201-302-7
NACRES:
NA.22

$94.40


Estimated to ship on30 May 2025


Request a Bulk Order

vapor pressure

2.9 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

222 °C

potency

835 mg/kg LD50, oral (Rat)

expl. lim.

1.0-7.5 % (v/v)

refractive index

n20/589 1.4659

pH

12.7 (25 °C, 10 g/L in H2O)

bp

162 °C/1013 hPa

mp

<-80 °C

transition temp

flash point 50 °C

density

0.92 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(C)(C)C(=N)N(C)C

InChI

1S/C5H13N3/c1-7(2)5(6)8(3)4/h6H,1-4H3

InChI key

KYVBNYUBXIEUFW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.915 - 0.917
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service