Skip to Content
Merck
All Photos(1)

Key Documents

8.21862

Sigma-Aldrich

Epoxycyclohexane

for synthesis

Synonym(s):

Epoxycyclohexane, 7-Oxabicyclo[4.1.0]heptane, Cyclohexene oxide

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$46.40
250 ML
$97.00

$46.40


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
50 ML
$46.40
250 ML
$97.00

About This Item

Empirical Formula (Hill Notation):
C6H10O
CAS Number:
Molecular Weight:
98.14
MDL number:
UNSPSC Code:
12352005
EC Index Number:
206-007-7
NACRES:
NA.22

$46.40


Estimated to ship on02 June 2025


Request a Bulk Order

vapor pressure

12 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

335 °C

potency

1060 mg/kg LD50, oral (Rat)
612 mg/kg LD50, skin (Rabbit)

expl. lim.

1.2-6.5 % (v/v)

bp

131-132 °C/1013 hPa

mp

-27 °C

transition temp

flash point 24 °C

density

0.97 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O1C2C1CCCC2

InChI

1S/C6H10O/c1-2-4-6-5(3-1)7-6/h5-6H,1-4H2

InChI key

ZWAJLVLEBYIOTI-UHFFFAOYSA-N

Application

  • Enantioselective Sensing for Catalyst Screening: A study discussed the use of epoxycyclohexane in the development of enantioselective sensing approaches, demonstrating its application in fluorous phase for high-throughput catalyst screening, crucial for pharmaceutical and fine chemical industries (Wu et al., 2021).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.972 - 0.973
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service