Skip to Content
Merck
All Photos(1)

Key Documents

8.21861

Sigma-Aldrich

1-Octanethiol

for synthesis

Synonym(s):

1-Octanethiol, n-Octyl mercaptan

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$47.90
500 ML
$102.00

$47.90


Estimated to ship on27 March 2025



Select a Size

Change View
100 ML
$47.90
500 ML
$102.00

About This Item

Linear Formula:
CH3(CH2)7SH
CAS Number:
Molecular Weight:
146.29
MDL number:
UNSPSC Code:
12352105
EC Index Number:
203-918-1
NACRES:
NA.22
Assay:
≥98.0% (GC)

$47.90


Estimated to ship on27 March 2025


vapor pressure

0.3 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

autoignition temp.

240 °C

potency

2000-2450 mg/kg LD50, oral (Rat)

bp

199 °C/1013 hPa

mp

-49 °C

transition temp

flash point 70 °C

solubility

0.004 g/L

density

0.84 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

SCCCCCCCC

InChI

1S/C8H18S/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3

InChI key

KZCOBXFFBQJQHH-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.841 - 0.845
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service