Skip to Content
Merck
All Photos(1)

Key Documents

8.21718

Sigma-Aldrich

Butyronitrile

for synthesis

Synonym(s):

Butyronitrile, Propyl cyanide, Butyric acid nitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7N
CAS Number:
Molecular Weight:
69.11
MDL number:
UNSPSC Code:
12352117
EC Index Number:
203-700-6
Pricing and availability is not currently available.

vapor pressure

19.7 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

488 °C

potency

50-100 mg/kg LD50, oral (Rat)
397 mg/kg LD50, skin (Rabbit)

expl. lim.

1.6 % (v/v)

bp

117-118 °C/1013 hPa

mp

-112 °C

transition temp

flash point 18 °C

solubility

33 g/L

density

0.79 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H7N/c1-2-3-4-5/h2-3H2,1H3

InChI key

KVNRLNFWIYMESJ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.790 - 0.792
Identity (IR): passes test

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service