Skip to Content
Merck
All Photos(1)

Key Documents

8.21240

Sigma-Aldrich

4-Methoxyphenylacetic acid

for synthesis

Synonym(s):

4-Methoxyphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O3
CAS Number:
Molecular Weight:
166.17
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-166-4
Pricing and availability is not currently available.

Assay

≥98.0% (acidimetric)

form

powder

bp

138-140 °C/4 hPa

mp

86-88 °C

transition temp

flash point 193 °C

bulk density

440 kg/m3

storage temp.

2-30°C

InChI

1S/C9H10O3/c1-12-8-4-2-7(3-5-8)6-9(10)11/h2-5H,6H2,1H3,(H,10,11)

InChI key

NRPFNQUDKRYCNX-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Melting range (lower value): ≥ 83 °C
Melting range (upper value): ≤ 86 °C
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

379.4 °F

Flash Point(C)

193 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service